2次元層状ペロブスカイト材料
溶液処理可能な材料
近年、溶液処理技術を用いて単層2次元ペロブスカイトを作製できることが発見され1、その後、発光ダイオード(LED)2、フォトトランジスタ3、太陽電池4,6、レーザー5などの2次元ペロブスカイトの光電子応用に関する研究が盛んに行われています。
直接および調整可能なバンドギャップ
フォトルミネッセンス2Dペロブスカイトは、層の厚さと、アミンやハロゲン化物の選択に応じて変化する狭い発光ピークを有する直接バンドギャップを持っています。フォトルミネッセンスベースのデバイス用に、最も一般的な2Dペロブスカイト組成の材料を取り揃えております。
水分安定性の向上
2Dペロブスカイトで作製した太陽電池は、3Dペロブスカイトと比較して湿った空気中での安定性が改善されています。
| Formula | Cat. No. | Layer Thickness | (RNH3)2(MeNH3)n−1 PbnX3n+1 | ||
|---|---|---|---|---|---|
| R | X | n | |||
| (BA)2PbI4 | 910961 | n=1 | Bu | I | 1 |
| (BA)2PbBr4 | 910953 | n=1 | Bu | Br | 1 |
| (PEA)2PbI4 | 910937 | n=1 | PE | I | 1 |
| (PEA)2PbBr4 | 910945 | n=1 | PE | Br | 1 |
| (BA)2MA)Pb2I7 | 912816 | n=2 | Bu | I | 2 |
| (BA)2(MA)2Pb3I10 | 912557 | n=3 | Bu | I | 3 |
| (BA)2(MA)3Pb4I13 | 914363 | n=4 | Bu | I | 4 |
| (BA)2(MA)4Pb5I16 | 912301 | n=5 | Bu | I | 5 |
申し訳ございませんが、想定外のエラーが発生しました。
Response not successful: Received status code 500
References
1.
Dou L, Wong AB, Yu Y, Lai M, Kornienko N, Eaton SW, Fu A, Bischak CG, Ma J, Ding T, et al. 2015. Atomically thin two-dimensional organic-inorganic hybrid perovskites. Science. 349(6255):1518-1521. https://doi.org/10.1126/science.aac7660
2.
Yuan M, Quan LN, Comin R, Walters G, Sabatini R, Voznyy O, Hoogland S, Zhao Y, Beauregard EM, Kanjanaboos P, et al. 2016. Perovskite energy funnels for efficient light-emitting diodes. Nature Nanotech. 11(10):872-877. https://doi.org/10.1038/nnano.2016.110
3.
Shao Y, Liu Y, Chen X, Chen C, Sarpkaya I, Chen Z, Fang Y, Kong J, Watanabe K, Taniguchi T, et al. 2017. Stable Graphene-Two-Dimensional Multiphase Perovskite Heterostructure Phototransistors with High Gain. Nano Lett.. 17(12):7330-7338. https://doi.org/10.1021/acs.nanolett.7b02980
4.
Cao DH, Stoumpos CC, Farha OK, Hupp JT, Kanatzidis MG. 2015. 2D Homologous Perovskites as Light-Absorbing Materials for Solar Cell Applications. J. Am. Chem. Soc.. 137(24):7843-7850. https://doi.org/10.1021/jacs.5b03796
5.
Raghavan CM, Chen T, Li S, Chen W, Lo C, Liao Y, Haider G, Lin C, Chen C, Sankar R, et al. 2018. Low-Threshold Lasing from 2D Homologous Organic–Inorganic Hybrid Ruddlesden–Popper Perovskite Single Crystals. Nano Lett.. 18(5):3221-3228. https://doi.org/10.1021/acs.nanolett.8b00990
6.
Hossain RF, Min M, Ma L, Sakri SR, Kaul AB. 2021. Carrier photodynamics in 2D perovskites with solution-processed silver and graphene contacts for bendable optoelectronics. npj 2D Mater Appl. 5(1): https://doi.org/10.1038/s41699-021-00214-3