Merck
ホーム組織工学GelMA(ゼラチンメタクリロイル)
GelMA(ゼラチンメタクリロイル)
Ali Khademhosseini教授(Harvard-MIT Division of Health Sciences and Technology, 米国)から、組織工学研究用ゼラチンメタクリロイル(900496など)の製品化のご提案をいただきました。
GelMa(ゼラチンメタクリロイル)は、天然高分子ゼラチンに光架橋可能なメタクリロイル基を導入した化合物で、印刷後に紫外または可視光で3D構造をゲル化することができます。所望の特徴(酵素分解性、調節可能な機械的および生物学的特性など)を有する複合ヒドロゲルプラットフォームを作製するために、PEG1,2、カーボンナノチューブ2、またはその他多くの物質と組み合わせることができる汎用的な材料です。また、3Dバイオプリンティングにおける光重合性ゼラチンメタクリロイルの使用4-7が報告されており、機能的血管ネットワークの形成6や内皮細胞形態の制御8、および心筋細胞の整列9をサポートします。さらに、ゼラチンメタクリロイルは、マイクロスフェア10,11やヒドロゲル12の形態での薬物送達用途にも検討されています。
上記応用例をはじめとする組織工学分野では、細孔径、分解速度、膨潤率などのハイドロゲル特性が要求されます。ゲル化後の機械的強度や剛柔特性に合わせて、様々なブルーム強度や官能基化度を持つGelMaをご用意しています。
3Dバイオプリンティング・3Dバイオインクに関するレビュー、プロトコル、関連製品紹介など、より詳細な情報については、「3Dバイオプリンティングハンドブック」をご覧ください。さらに、Ali教授によるウェビナー「再生工学用ナノ・マイクロ加工ハイドロゲル(日本語字幕付き)」も併せてご参考ください。
GelMAの構造式
関連製品
申し訳ございませんが、想定外のエラーが発生しました。
Response not successful: Received status code 500
Material Matters(季刊誌)
Material Matters(季刊誌)
Material Mattersは、材料科学分野に関する季刊ニュースレターです。世界各国の第一線の研究者によるテクニカルレビューを掲載しています。
ChemNews(ケムニュース)
ChemNews(ケムニュース)
有機合成化学・材料科学分野の研究者の方に新製品情報をいち早くお届けするメールニュースです。
構造式カタログ
構造式カタログ
10万点以上のシグマアルドリッチ製品の構造式を一覧表示できます。カテゴリや製品英名で絞込み検索が可能です。
References
1.
Hutson CB, Nichol JW, Aubin H, Bae H, Yamanlar S, Al-Haque S, Koshy ST, Khademhosseini A. 2011. Synthesis and Characterization of Tunable Poly(Ethylene Glycol): Gelatin Methacrylate Composite Hydrogels. Tissue Engineering Part A. 17(13-14):1713-1723. https://doi.org/10.1089/ten.tea.2010.0666
2.
Nichol JW, Koshy ST, Bae H, Hwang CM, Yamanlar S, Khademhosseini A. 2010. Cell-laden microengineered gelatin methacrylate hydrogels. Biomaterials. 31(21):5536-5544. https://doi.org/10.1016/j.biomaterials.2010.03.064
3.
Shin SR, Jung SM, Zalabany M, Kim K, Zorlutuna P, Kim Sb, Nikkhah M, Khabiry M, Azize M, Kong J, et al. 2013. Carbon-Nanotube-Embedded Hydrogel Sheets for Engineering Cardiac Constructs and Bioactuators. ACS Nano. 7(3):2369-2380. https://doi.org/10.1021/nn305559j
4.
Bertassoni LE, Cardoso JC, Manoharan V, Cristino AL, Bhise NS, Araujo WA, Zorlutuna P, Vrana NE, Ghaemmaghami AM, Dokmeci MR, et al. Direct-write bioprinting of cell-laden methacrylated gelatin hydrogels. Biofabrication. 6(2):024105. https://doi.org/10.1088/1758-5082/6/2/024105
5.
Billiet T, Gevaert E, De Schryver T, Cornelissen M, Dubruel P. 2014. The 3D printing of gelatin methacrylamide cell-laden tissue-engineered constructs with high cell viability. Biomaterials. 35(1):49-62. https://doi.org/10.1016/j.biomaterials.2013.09.078
6.
Skardal A, Zhang J, McCoard L, Xu X, Oottamasathien S, Prestwich GD. 2010. Photocrosslinkable Hyaluronan-Gelatin Hydrogels for Two-Step Bioprinting. Tissue Engineering Part A. 16(8):2675-2685. https://doi.org/10.1089/ten.tea.2009.0798
7.
Boere KW, Visser J, Seyednejad H, Rahimian S, Gawlitta D, van Steenbergen MJ, Dhert WJ, Hennink WE, Vermonden T, Malda J. 2014. Covalent attachment of a three-dimensionally printed thermoplast to a gelatin hydrogel for mechanically enhanced cartilage constructs. Acta Biomaterialia. 10(6):2602-2611. https://doi.org/10.1016/j.actbio.2014.02.041
8.
Nikkhah M, Eshak N, Zorlutuna P, Annabi N, Castello M, Kim K, Dolatshahi-Pirouz A, Edalat F, Bae H, Yang Y, et al. 2012. Directed endothelial cell morphogenesis in micropatterned gelatin methacrylate hydrogels. Biomaterials. 33(35):9009-9018. https://doi.org/10.1016/j.biomaterials.2012.08.068
9.
Tsang KMC, Annabi N, Ercole F, Zhou K, Karst DJ, Li F, Haynes JM, Evans RA, Thissen H, Khademhosseini A, et al. 2015. Facile One-Step Micropatterning Using Photodegradable Gelatin Hydrogels for Improved Cardiomyocyte Organization and Alignment. Adv. Funct. Mater.. 25(6):977-986. https://doi.org/10.1002/adfm.201403124
10.
Cha C, Oh J, Kim K, Qiu Y, Joh M, Shin SR, Wang X, Camci-Unal G, Wan K, Liao R, et al. 2014. Microfluidics-Assisted Fabrication of Gelatin-Silica Core–Shell Microgels for Injectable Tissue Constructs. Biomacromolecules. 15(1):283-290. https://doi.org/10.1021/bm401533y
11.
Nguyen AH, McKinney J, Miller T, Bongiorno T, McDevitt TC. 2015. Gelatin methacrylate microspheres for controlled growth factor release. Acta Biomaterialia. 13101-110. https://doi.org/10.1016/j.actbio.2014.11.028
12.
Shivakumar H, Gupta N, Satish C. 2007. Preparation and characterization of gelatin-poly(methacrylic acid) interpenetrating polymeric network hydrogels as a ph-sensitive delivery system for glipizide. Indian J Pharm Sci. 69(1):64. https://doi.org/10.4103/0250-474x.32110