Merck
  • Behavioral effects of nicotinic antagonist mecamylamine in a rat model of depression: prefrontal cortex level of BDNF protein and monoaminergic neurotransmitters.
Behavioral effects of nicotinic antagonist mecamylamine in a rat model of depression: prefrontal cortex level of BDNF protein and monoaminergic neurotransmitters.
Psychopharmacology (2014-10-16)
Sawsan Aboul-Fotouh
RESUMEN
Several studies have pointed to the nicotinic acetylcholine receptor (nAChR) antagonists, such as mecamylamine (MEC), as a potential therapeutic target for the treatment of depression. The present study evaluated the behavioral and neurochemical effects of chronic administration of MEC (1, 2, and 4 mg/kg/day, intraperitoneally (i.p.)) in Wistar rats exposed to chronic restraint stress (CRS, 4 h × 6 W). MEC prevented CRS-induced depressive-like behavior via increasing sucrose preference, body weight, and forced swim test (FST) struggling and swimming while reducing immobility in FST and hypothalamic-pituitary-adrenal (HPA) axis hyperactivity (adrenal gland weight and serum corticosterone). At the same time, MEC amended CRS-induced anxiety as indicated by decreasing central zone duration in open field test and increasing active interaction duration. Additionally, MEC modulated the prefrontal cortex (PFC) level of brain-derived neurotrophic factor (BDNF), 5-hydroxy tryptamine (5-HT), and norepinephrine (NE). In conclusion, the present data suggest that MEC possesses antidepressant and anxiolytic-like activities in rats exposed to CRS. These behavioral effects may be in part mediated by reducing HPA axis hyperactivity and increasing PFC level of BDNF and monoamines. Accordingly, these findings further support the hypothesis that nAChRs blockade might afford a novel promising strategy for pharmacotherapy of depression.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido perclórico, ACS reagent, 70%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido L-ascórbico, powder, suitable for cell culture, γ-irradiated
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido L-ascórbico, BioXtra, ≥99.0%, crystalline
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido L-ascórbico, suitable for cell culture, suitable for plant cell culture, ≥98%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido L-ascórbico, 99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido L-ascórbico, reagent grade, crystalline
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Ácido L-ascórbico, Pharmaceutical Secondary Standard; Certified Reference Material
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido perclórico, 70%, 99.999% trace metals basis
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido perclórico, ACS reagent, 60%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
En este momento no podemos mostrarle ni los precios ni la disponibilidad
USP
Ácido L-ascórbico, United States Pharmacopeia (USP) Reference Standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido L-ascórbico, ACS reagent, ≥99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Ácido L-ascórbico, analytical standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido perclórico, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 70.0-72.0%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido L-ascórbico, meets USP testing specifications
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido L-ascórbico, reagent grade
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido L-ascórbico, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido L-ascórbico, FCC, FG
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Mycophenolic acid, ≥98%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido L-ascórbico, BioUltra, ≥99.5% (RT)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Ácido L-ascórbico, European Pharmacopoeia (EP) Reference Standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Mecamylamine hydrochloride
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido L-ascórbico, puriss. p.a., ≥99.0% (RT)
En este momento no podemos mostrarle ni los precios ni la disponibilidad