Merck
Deficient histone H3 propionylation by BRPF1-KAT6 complexes in neurodevelopmental disorders and cancer.
Science advances (2020-02-06)
Kezhi Yan, Justine Rousseau, Keren Machol, Laura A Cross, Katherine E Agre, Cynthia Forster Gibson, Anne Goverde, Kendra L Engleman, Hannah Verdin, Elfride De Baere, Lorraine Potocki, Dihong Zhou, Maxime Cadieux-Dion, Gary A Bellus, Monisa D Wagner, Rebecca J Hale, Natacha Esber, Alan F Riley, Benjamin D Solomon, Megan T Cho, Kirsty McWalter, Roy Eyal, Meagan K Hainlen, Bryce A Mendelsohn, Hillary M Porter, Brendan C Lanpher, Andrea M Lewis, Juliann Savatt, Isabelle Thiffault, Bert Callewaert, Philippe M Campeau, Xiang-Jiao Yang
ABSTRAKT
Lysine acetyltransferase 6A (KAT6A) and its paralog KAT6B form stoichiometric complexes with bromodomain- and PHD finger-containing protein 1 (BRPF1) for acetylation of histone H3 at lysine 23 (H3K23). We report that these complexes also catalyze H3K23 propionylation in vitro and in vivo. Immunofluorescence microscopy and ATAC-See revealed the association of this modification with active chromatin. Brpf1 deletion obliterates the acylation in mouse embryos and fibroblasts. Moreover, we identify BRPF1 variants in 12 previously unidentified cases of syndromic intellectual disability and demonstrate that these cases and known BRPF1 variants impair H3K23 propionylation. Cardiac anomalies are present in a subset of the cases. H3K23 acylation is also impaired by cancer-derived somatic BRPF1 mutations. Valproate, vorinostat, propionate and butyrate promote H3K23 acylation. These results reveal the dual functionality of BRPF1-KAT6 complexes, shed light on mechanisms underlying related developmental disorders and various cancers, and suggest mutation-based therapy for medical conditions with deficient histone acylation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Butyryl coenzyme A lithium salt hydrate, ≥90%
Sigma-Aldrich
Sodium propionate, ≥99.0%, BioReagent, suitable for insect cell culture
Sigma-Aldrich
n-Propionyl coenzyme A lithium salt, ≥85%
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Przeciwciało przeciwko acetylo-histonie H3 (Lys14), serum, Upstate®
Sigma-Aldrich
Crotonoyl coenzyme A trilithium salt, ~90% (HPLC)
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
DL-β-Hydroxybutyric acid sodium salt, ~98%
Sigma-Aldrich
Valproic acid sodium salt, 98%
Sigma-Aldrich
Acetyl coenzyme A trisodium salt, ≥93% (HPLC), powder
Millipore
FLAG® Peptide, lyophilized powder
Supelco
4-Hydroxytamoxifen, analytical standard, (E) and (Z) isomers (50:50)
Sigma-Aldrich
SAHA, ≥98% (HPLC)
Sigma-Aldrich
Przeciwciało przeciwko acetylo-histonie H3 (Lys18), serum, Upstate®
Sigma-Aldrich
Przeciwciało przeciwko acetylo-histonie H3 (Lys23), serum, Upstate®