Merck
Identification of the toxic trigger in mushroom poisoning.
Nature chemical biology (2009-05-26)
Masanori Matsuura, Yoko Saikawa, Kosei Inui, Koichi Nakae, Masayuki Igarashi, Kimiko Hashimoto, Masaya Nakata
ABSTRAKT
We have isolated the small, highly strained carboxylic acid cycloprop-2-ene carboxylic acid from the Asian toxic mushroom Russula subnigricans. This compound is responsible for fatal rhabdomyolysis, a new type of mushroom poisoning that is indicated by an increase in serum creatine phosphokinase activity in mice. We found that polymerization of the compound at high concentrations via ene reaction abolishes its toxicity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propiolic acid, 95%
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Butyric acid, natural, ≥99%, FCC, FG
Supelco
Butyric acid, analytical standard
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Butyric acid, ≥99%, FG
Sigma-Aldrich
Butyric acid, ≥99%
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Crotonic acid, 98%
Supelco
Propionic acid, analytical standard
Sigma-Aldrich
Propionic acid, ≥99.5%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
p-Phenylenediamine, ≥99.0% (GC/NT)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
p-Phenylenediamine, 98% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%