Merck
How well can the Caco-2/Madin-Darby canine kidney models predict effective human jejunal permeability?
Journal of medicinal chemistry (2010-04-09)
Alex Avdeef, Kin Y Tam
ABSTRAKT
The study aimed to predict effective human jejunal permeability (P(eff)) using a biophysical model based on parametrized paracellular, aqueous boundary layer, and transcellular permeabilities, and the villus-fold surface area expansion factor (k(VF)). Published human jejunal data (119 P(eff), 53 compounds) were analyzed by a regression procedure incorporating a dual-pore size paracellular model. Transcellular permeability, scaled by k(VF), was equated to that of Caco-2 at pH 6.5. The biophysical model predicted human jejunal permeability data within the experimental uncertainty. This investigation revealed several surprising predictions: (i) many molecules permeate predominantly (but not exclusively) by the paracellular route, (ii) the aqueous boundary layer thickness in the intestinal perfusion experiments is larger than expected, (iii) the mucosal surface area in awake humans is apparently nearly entirely accessible to drug absorption, and (iv) the relative "leakiness" of the human jejunum is not so different from that observed in a number of published Caco-2 studies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Supelco
Ibuprofen
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Sigma-Aldrich
Griseofulvin, from Penicillium griseofulvum, 97.0-102.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, Molecular Biology, ≥95%
Sigma-Aldrich
Glipizide, solid
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%