Merck
Novel EDA p.Ile260Ser mutation linked to non-syndromic hypodontia.
Journal of dental research (2013-04-30)
Y Yang, L Luo, J Xu, P Zhu, W Xue, J Wang, W Li, M Wang, K Cheng, S Liu, Z Tang, B Z Ring, L Su
ABSTRAKT
Hypodontia, a tooth developmental disease, can affect chewing and pronunciation. Mutations in the ectodysplasin-A (EDA) gene can lead to both X-linked hypohidrotic ectodermal dysplasia (XLHED) and non-syndromic hypodontia (NSH). However, the mechanism by which these 2 related but different disorders are caused by the distinct mutations in EDA is unknown. In this study, we identified a novel missense mutation (c.779 T>G) in a Chinese family with NSH via a direct sequencing approach. This mutation results in an Ile260Ser substitution in the tumor necrosis factor (TNF) homology domain. Homology modeling suggests that this alteration may induce a conformational change in the hydrophobic center of the TNF homology domain. Furthermore, by exploring systematic 3D conformation analysis and calculation of residue relative solvent accessibility (RSA) for all the reported mutated amino acid sites on EDA's TNF homology domain, we found that the site mutations at the interior may be linked to XLHED, while those at the surface are more likely to be associated with NSH. These findings may aid in the discovery of unidentified functionally significant mutation sites in the EDA gene and provide a new way to clarify the mechanisms by which the XLHED and NSH phenotypes arise from mutations in the same gene.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Guanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
Guanine, 98%
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Thymine, BioReagent, suitable for cell culture
Sigma-Aldrich
Guanine, BioUltra
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
SAFC
L-Valine
Sigma-Aldrich
Thymine, ≥99%
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Serine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
L-Isoleucine
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland