Merck
Vitamins - wrong approaches.
International journal for vitamin and nutrition research. Internationale Zeitschrift fur Vitamin- und Ernahrungsforschung. Journal international de vitaminologie et de nutrition (2013-06-26)
Ulrich Moser
ABSTRAKT
Deficiencies of essential nutrients have been responsible for many epidemic outbreaks of deficiency diseases in the past. Large observational studies point at possible links between nutrition and chronic diseases. Low intake of antioxidant vitamins e. g. have been correlated to increased risk of cardiovascular diseases or cancer. The main results of these studies are indications that an intake below the recommendation could be one of the risk factors for chronic diseases. There was hardly any evidence that amounts above the RDA could be of additional benefit. Since observational studies cannot prove causality, the scientific community has been asking for placebo-controlled, randomized intervention trials (RCTs). Thus, the consequences of the epidemiological studies would have been to select volunteers whose baseline vitamin levels were below the recommended values. The hypothesis of the trial should be that correcting this risk factor up to RDA levels lowers the risk of a disease like CVD by 20 - 30 %. However, none of the RCTs of western countries was designed to correct a chronic marginal deficiency, but they rather tested whether an additional supplement on top of the recommended values would be beneficial in reducing a disease risk or its prognosis. It was, therefore, not surprising that the results were disappointing. As a matter of fact, the results confirmed the findings of the observational studies: chronic diseases are the product of several risk factors, among them most probably a chronic vitamin deficiency. Vitamin supplements could only correct the part of the overall risk that is due to the insufficient vitamin intake.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
β-Carotene, synthetic, ≥95% (HPLC), crystalline
Sigma-Aldrich
β-Carotene, synthetic, ≥93% (UV), powder
Supelco
β-Carotene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard