Merck
[Role of serine/threonine and tyrosine protein phosphatases in command Helix lucorum neurons at the cellular correlate of habituation].
Zhurnal vysshei nervnoi deiatelnosti imeni I P Pavlova (2013-07-23)
A S Pivovarov, G B Murzina, M S Tret'iakova, D A Makhnovskiĭ
ABSTRAKT
Effects of some inhibitors of serine/threonine and tyrosine protein phosphatases on the depression and spontaneous recovery of the acetylcholine-induced inward current (ACh-current) in command Helix neurons of defensive behavior at the cellular correlate of habituation were investigated. The following drugs were used: okadaic acid (reduces activity ofphosphatases PP1 and PP2A), endothall (PP2A), cyclosporine A and cypermethrin (PP2B), CCT007093 (PPM1D), dephostatin (blocks tyrosine phosphatases). All used inhibitors modify the depression flow, and endothall reduces spontaneous recovery of ACh-current also. Obtained results indicate that changes in cholinosensitivity of command neurons depend on activity of all investigated protein phosphatases. Mathematical model considers the possibility of different localizations of receptors in a neuron and regularity of transitions between them. This model makes it possible to conclude participation indicated phosphatases in mobility of membrane cholinoreceptors ensuring the ACh-current modification at the cellular correlate of habituations. Comparison of experimental and calculated curves of ACh-current change allows to conclude that the main target of protein phosphatases is the transport system of a neuron--cytoskeleton and motor proteins.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Cyclosporine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetylcholine chloride, pkg of 150 mg (per vial)
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC)
Sigma-Aldrich
L-Serine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
Cyclosporin A solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetylcholine chloride, suitable for cell culture
Supelco
Cypermethrin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Cypermethrin, PESTANAL®, analytical standard
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC), free-flowing, Redi-Dri
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
SAFC
L-Threonine
Sigma-Aldrich
Okadaic acid from Prorocentrum concavum, 92-100% (HPLC)
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, Molecular Biology, ≥95%
Sigma-Aldrich
Acetylcholine bromide, ≥99%
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
Acetylcholine iodide, ≥97%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
α-Cypermethrin, PESTANAL®, analytical standard
Supelco
Cypermethrin solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material