Merck
Heparin-induced release of protein-bound solutes during hemodialysis is an in vitro artifact.
Clinical chemistry (2001-04-28)
R De Smet, J Van Kaer, H Liebich, G Lesaffer, A Verstraete, A Dhondt, P Duym, N Lameire, R Vanholder
ABSTRAKT
Several studies have pointed to a release of drugs or protein-bound solutes from their binding sites during heparinization. The effect is attributed to the metabolism of triglycerides to free fatty acids (FFAs), which compete with drugs for protein binding sites. This study evaluated the impact of intradialytic heparin on the free concentration of the uremic toxin p-cresol and on FFAS: Blood samples from hemodialysis (HD) patients, before and during HD, were collected with selected anticoagulation strategies. We assessed the effects of standing time, temperature, pH, and the addition of a lipase inhibitor, tetrahydrolipstatin (THL) to blood samples on the free p-cresol concentration. p-Cresol was analyzed by HPLC with fluorescence detection. We measured FFAs by gas chromatography, and the free fractions of added valproic acid and phenytoin were evaluated by fluorescence polarization immunoassay. In blood samples (n = 22) not submitted to a specific treatment, free p-cresol increased from 9.9 +/- 5.1 to 31.9 +/- 22.3 micromol/L after 30 min of heparin HD (P < 0.001) and correlated significantly with FFAs (r = 0.80; P = 0.002; n = 12). There was no increase in free p-cresol during heparin-free HD (n = 6) and trisodium citrate HD (n = 9). In addition, p-cresol in ultrafiltrates (n = 3) did not correspond to the free p-cresol in heparinized blood, suggesting that the increase in free p-cresol was artifactual. The release of p-cresol in the test tube was enhanced by standing time (n = 6), sample temperature (n = 6), and alkaline pH (n = 6). Inhibition of lipase activity with THL prevented the increase of FFAs (n = 6) and the release of free p-cresol during HD (n = 22). These results were corroborated by the study of the free fraction of valproic acid (n = 6) and phenytoin (n = 6). The free concentrations of protein-bound solutes in plasma of heparinized patients are influenced by external factors that alter the lipase activity in the test tube. The free fraction does not increase during HD when lipase activity is neutralized at the time of blood sampling, so that previously reported increases are probably artifacts.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
Zinc, purum, powder
Sigma-Aldrich
Zinc, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, granular, 30-100 mesh, 99%
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
DL-Homocysteine, ≥95% (titration)
Supelco
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Supelco
p-Cresol, analytical standard
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
trans-4-Hydroxy-L-proline, ≥99%
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
p-Cresol, ≥99%, FG