Merck
A new self-microemulsifying mouth dissolving film to improve the oral bioavailability of poorly water soluble drugs.
Drug development and industrial pharmacy (2012-09-27)
Lu Xiao, Tao Yi, Ying Liu
ABSTRAKT
A new self-microemulsifying mouth dissolving film (SMMDF) for poorly water-soluble drugs such as indomethacin was developed by incorporating self-microemulsifying components with solid carriers mainly containing microcrystalline cellulose, low-substituted hydroxypropyl cellulose and hypromellose. The uniformity of dosage units of the preparation was acceptable according to the criteria of Chinese Pharmacopoeia 2010. The SMMDF was disintegrated within 20 s after immersion into water, released completely at 5 min in the dissolution medium and achieved microemulsion particle size of 28.81 ± 3.26 nm, which was similar to that of liquid self- microemulsifying drug delivery system (SMEDDS). Solid state characterization of the SMMDF was performed by SEM, DSC and X-ray powder diffraction. Results demonstrated that indomethacin in the SMMDF was in the amorphous state, which might be due to self-microemulsifying ingredients. Pharmacokinetic parameters in rats including T(max), C(max), AUC were similar between the SMMDF and liquid SMEDDS. AUC and C(max) from the SMMDF were significantly higher than those from the common mouth dissolving film or the conventional tablet, and Tmax from SMMDF group was also significantly decreased. These findings suggest that the SMMDF is a new promising dosage form, showing notable characteristics of convenience, quick onset of action and enhanced oral bioavailability of poorly water-soluble drugs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methyl cellulose, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
Hydroxypropyl cellulose, average Mw ~1,000,000, powder, 20 mesh particle size (99% through)
Sigma-Aldrich
Hydroxypropyl cellulose, average Mw ~100,000, powder, 20 mesh particle size (99% through)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
Hydroxypropyl cellulose, average Mw ~370,000, powder, 20 mesh particle size (99% through)
Sigma-Aldrich
Hydroxypropyl cellulose, average Mw ~80,000, powder, 20 mesh particle size (99% through)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.