Merck
  • Evaluation of cycloserine-cefoxitin fructose agar (CCFA), CCFA with horse blood and taurocholate, and cycloserine-cefoxitin mannitol broth with taurocholate and lysozyme for recovery of Clostridium difficile isolates from fecal samples.
Evaluation of cycloserine-cefoxitin fructose agar (CCFA), CCFA with horse blood and taurocholate, and cycloserine-cefoxitin mannitol broth with taurocholate and lysozyme for recovery of Clostridium difficile isolates from fecal samples.
Journal of clinical microbiology (2013-06-28)
Kerin L Tyrrell, Diane M Citron, Eliza S Leoncio, C Vreni Merriam, Ellie J C Goldstein
ABSTRAKT
Cycloserine-cefoxitin fructose agar (CCFA), CCFA with horse blood and taurocholate (CCFA-HT), and cycloserine-cefoxitin mannitol broth with taurocholate and lysozyme (CCMB-TAL) were compared for recovery of Clostridium difficile from 120 stool specimens. Compared to CCFA, CCFA-HT enhanced C. difficile growth and improved recovery by 4%. In a separate study, 9% (8/91) of stool samples previously C. difficile negative on plate medium were C. difficile positive when cultured in CCMB-TAL.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Millipore
Agar, high purity, low ionic content, low gel strenght, suitable for microbiology
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
Noble agar
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Sigma-Aldrich
Select agar
Sodium taurocholate, BRP, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Agar, ash 2.0-4.5%
Millipore
Agar, suitable for microbiology
Millipore
Agar, powder, suitable for microbiology
Cefoxitin sodium, European Pharmacopoeia (EP) Reference Standard
Supelco
Cefoxitin sodium salt
Mannitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Cycloserine
Sigma-Aldrich
D-Cycloserine, synthetic