Merck
[Peculiarity of reassortment of current wild type influenza viruses with master donor viruses for live influenza vaccine].
Voprosy virusologii (2014-03-20)
I V Kiseleva, E A Bazhenova, N V Larionova, E A Fedorova, I A Dubrovina, I N Isakova-Sivak, L G Rudenko
ABSTRAKT
The live attenuated influenza vaccine (LAIV) currently licensed in Russia consists of the reassortant viruses with hemagglutinin (HA) and neuraminidase (NA) gene segments from the circulating wild-type viruses and the six internal protein-encoding gene segments from cold-adapted master donor viruses (MDV) A/Leningrad/134/17/57 (H2N2) or B/USSR/60/69. Presently, only classical reassortment technique is approved for the generation of Russian LAIV strains. In this work, we describe the obstacles to the development of LAIV 6:2 vaccine strains depending on the phenotypic properties of the wild-type viruses used for reassortment. It was demonstrated that the highest percentage of 6:2 vaccine reassortants could be achieved when wild-type parental viruses were resistant to non-specific gamma-inhibitors. It was shown that it was impossible to generate 6:2 vaccine reassortants possessing six internal genes of the AILeningrad113417/57 (H2N2) master donor virus and avian HA and NA genes from H5N1-PR8 viruses using classical reassortment technique. It was suggested that strong constellation effects between the gene segments of the parental viruses could affect the virus gene reassortment. A strong interaction between the genome segments encoding neuraminidase of avian origin and PB2 gene of PR8 virus was observed. When the PB2 gene was inherited from cold-adapted master donor virus, the neuraminidase was also found to be of MDV origin.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
α(2→3) Neuraminidase from Streptococcus pneumoniae, buffered aqueous solution
Sigma-Aldrich
Neuraminidase Agarose from Clostridium perfringens (C. welchii), Type VI-A, ammonium sulfate suspension
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VI, lyophilized powder, 6-15 units/mg protein (using 4MU-NANA), 2-10 units/mg protein (mucin)
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type X, lyophilized powder, ≥50 units/mg protein (using 4MU-NANA)
Sigma-Aldrich
α(2→3,6) Neuraminidase from Clostridium perfringens (C. welchii), recombinant, expressed in E. coli, buffered aqueous solution, ≥250 units/mg protein
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VIII, lyophilized powder, 10-20 units/mg protein (using 4MU-NANA), 3.5-8.0 units/mg protein (mucin)
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, Proteomics Grade, suitable for MALDI-TOF MS
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, ≥1.5 U/mL, specific activity ≥ 1.5U/mg protein
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type II, buffered aqueous solution, 8-24 units/mg protein (Lowry, using NAN-lactose)
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type III, buffered aqueous solution, 0.2 μm filtered, 1-5 units/mg protein (Lowry, using NAN-lactose)