Merck
HER2 mutation status in Japanese HER2-negative breast cancer patients.
Japanese journal of clinical oncology (2014-05-08)
Yumi Endo, Yu Dong, Nobuyasu Yoshimoto, Tomoko Asano, Yukari Hato, Hiroko Yamashita, Shinya Sato, Satoru Takahashi, Yoshitaka Fujii, Tatsuya Toyama
ABSTRAKT
Human epidermal growth factor receptor 2 (HER2) gene amplification is a major therapeutic target in breast cancer, and has been introduced as a predictive biomarker to identify patients who may benefit from therapy with anti-human epidermal growth factor receptor 2 agents. Human epidermal growth factor receptor 2 somatic mutations have been reported in patients without human epidermal growth factor receptor 2 gene amplification. Since these are activating mutations, these patients may also benefit from human epidermal growth factor receptor 2-targeted drugs. In this study, we searched for human epidermal growth factor receptor 2 mutations in a group of 286 Japanese breast cancer patients with human epidermal growth factor receptor 2-negative tumors. The activating mutations of human epidermal growth factor receptor 2 identified were analyzed by direct Sanger sequencing of two major areas: the extracellular domain at 309-310 and the kinase domain between 755 and 781. Two tumors were found to have a human epidermal growth factor receptor 2 somatic mutation; one with I767M mutation and another with D769Y. No mutation was observed in the extracellular domain. One of these patients with human epidermal growth factor receptor 2 mutation recurred early with liver metastasis. Better knowledge of human epidermal growth factor receptor 2 mutation status will help us to choose personalized molecular targeted therapy for use in human epidermal growth factor receptor 2-negative Japanese breast cancer patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
L-Methionine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)
SAFC
L-Methionine
SAFC
L-Isoleucine
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
Methionine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Methionine, Pharmaceutical Secondary Standard; Certified Reference Material
Tyrosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
SAFC
L-Aspartic acid
SAFC
L-Tyrosine
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland