Merck
Determination of aromatic amines in human urine using comprehensive multi-dimensional gas chromatography mass spectrometry (GCxGC-qMS).
Analytical and bioanalytical chemistry (2014-08-22)
Xolelwa Lamani, Simeon Horst, Thomas Zimmermann, Torsten C Schmidt
ABSTRAKT
Aromatic amines are an important class of harmful components of cigarette smoke. Nevertheless, only few of them have been reported to occur in urine, which raises questions on the fate of these compounds in the human body. Here we report on the results of a new analytical method, in situ derivatization solid phase microextraction (SPME) multi-dimensional gas chromatography mass spectrometry (GCxGC-qMS), that allows for a comprehensive fingerprint analysis of the substance class in complex matrices. Due to the high polarity of amino compounds, the complex urine matrix and prevalence of conjugated anilines, pretreatment steps such as acidic hydrolysis, liquid-liquid extraction (LLE), and derivatization of amines to their corresponding aromatic iodine compounds are necessary. Prior to detection, the derivatives were enriched by headspace SPME with the extraction efficiency of the SPME fiber ranging between 65 % and 85 %. The measurements were carried out in full scan mode with conservatively estimated limits of detection (LOD) in the range of several ng/L and relative standard deviation (RSD) less than 20 %. More than 150 aromatic amines have been identified in the urine of a smoking person, including alkylated and halogenated amines as well as substituted naphthylamines. Also in the urine of a non-smoker, a number of aromatic amines have been identified, which suggests that the detection of biomarkers in urine samples using a more comprehensive analysis as detailed in this report may be essential to complement the approach of the use of classic biomarkers.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets