Merck
  • Polymorphism of the multidrug resistance 1 gene MDR1/ABCB1 C3435T and response to antiepileptic drug treatment in temporal lobe epilepsy.
Polymorphism of the multidrug resistance 1 gene MDR1/ABCB1 C3435T and response to antiepileptic drug treatment in temporal lobe epilepsy.
Seizure (2014-12-03)
Ida Manna, Antonio Gambardella, Angelo Labate, Laura Mumoli, Edoardo Ferlazzo, Franco Pucci, Umberto Aguglia, Aldo Quattrone
ABSTRAKT
A synonymous C to T variant at position 3435 (c.3435C>T) is one common polymorphism of the multidrug resistant 1 (MDR1) gene, which encodes the major transmembrane efflux transporter P-glycoprotein. It has been suggested that this polymorphism, and more specifically the 3435CC genotype, may be associated with the response to antiepileptic drug treatment. Here we wished to examine the role of such a candidate variant in a cohort of 175 patients (98 women and 76 men; mean ± SD age: 47.90 ± 17.64) with temporal lobe epilepsy (TLE). Patients were classified according to whether they had drug-responsive (n=134) or drug-resistant (n=41) epilepsy. We also enrolled 175 healthy controls (93 women and 82 men; mean ± SD age: 72.5 ± 6.8), matched for sex and ethnicity. Patients and controls were genotyped for detection of the 3435C>T polymorphism, but the analysis showed no significant association between the CC genotype and the risk of drug-resistant epilepsy. These findings rule out the MDR1 c.3435C>T polymorphism having a major role or increasing the risk of drug-resistance suggesting a revision is required to determine the contribution of this polymorphism in predicting drug response in epilepsy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O