Merck
  • Genetic characterization of oropharyngeal trichomonad isolates from wild birds indicates that genotype is associated with host species, diet and presence of pathognomonic lesions.
Genetic characterization of oropharyngeal trichomonad isolates from wild birds indicates that genotype is associated with host species, diet and presence of pathognomonic lesions.
Avian pathology : journal of the W.V.P.A (2014-09-30)
M C Martínez-Herrero, J Sansano-Maestre, I López Márquez, E Obón, C Ponce, J González, M M Garijo-Toledo, M T Gómez-Muñoz
ABSTRAKT
Oropharyngeal trichomonad isolates of wild birds from Spain were studied. A total of 1688 samples (1214 of predator birds and 474 of prey species) from wildlife recovery centres and scientific bird-ringing campaigns were analysed from 2011 to 2013. The overall infection prevalence was 20.3% (11.4% in predator birds and 43.3% in prey species). Pathognomonic lesions were present in 26% of the infected birds (57.3% in predator birds and 4.9% in prey species). The most commonly parasitized species were the goshawk (Accipiter gentilis, 74.5%) and the rock pigeon (Columba livia, 79.4%). Host species in which the parasite has not been previously analysed by polymerase chain reaction and sequencing in Spain are also reported: Columba palumbus, Streptopelia turtur, Pica pica, A. gentilis, Accipiter nisus, Asio otus, Bubo bubo, Buteo buteo, Circus aeruginosus, Circus cyaneus, Falco naumanni, Falco peregrinus, Neophron percnopterus, Otus scops, Pernis apivorus and Strix aluco. Sequence analysis of the ITS1/5.8S/ITS2 region revealed five different genotypes and also some mixed infections. A relationship between genotype and host species was observed, but only two genotypes (ITS-OBT-Tg-1and ITS-OBT-Tg-2) were widely distributed. Genotype ITS-OBT-Tg-1 was most frequently found in predator birds and statistically associated with pathognomonic lesions. Non-strict ornithophagous species were at higher risk to develop disease than ornithophagous ones. Genotypes ITS-OBT-Tcl-1 and ITS-OBT-Tcl-2 are new reports, and ITS-OBT-Tvl-5 is reported for the first time in Spain. They showed higher genetic homology to Trichomonas canistomae and Trichomonas vaginalis than to Trichomonas gallinae, indicating the possibility of new species within this genus.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ceftiofur, VETRANAL®, analytical standard
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
SAFC
L-Cysteine
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Kwas askorbinowy, United States Pharmacopeia (USP) Reference Standard