Merck
Exopolysaccharide of Antarctic bacterium Pseudoaltermonas sp. S-5 induces apoptosis in K562 cells.
Carbohydrate polymers (2015-02-11)
Guochuang Chen, Wen Qian, Jing Li, Yanghui Xu, Kaoshan Chen
ABSTRAKT
The aim of this study was to investigate the anticancer activity of exopolysaccharide (PEP) of Antarctic bacterium Pseudoaltermonas sp. S-5 and elucidate the underlying molecular mechanisms. PEP significantly inhibited the growth of human leukemia K562 cells. Results of morphological characterization showed that PEP-treated cells displayed typical morphological characteristics of apoptosis such as condensation of chromatin and formation of apoptotic bodies. Flow cytometry analyses and colorimetric assay demonstrated that PEP induced collapse of mitochondrial membrane potential and activation of caspase-9, which indicated that intrinsic apoptotic signaling pathway was involved in apoptosis induced by PEP in K562 cells. Western blot analysis showed that PEP increased the ratio of Bax/Bcl-2. In addition, calcium signal might contribute to the cytotoxicity of PEP against K562 cells. These findings suggest that PEP may be potentially effective drug against human leukemia.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
1,2-Bis(2-amino-5-methylphenoxy)ethane-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, ≥95% (HPLC)
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99%
Millipore
D-(+)-Mannose, ≥99.0% (sum of enantiomers, HPLC), suitable for microbiology
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Dioctan 2′,7′-dichlorodihydrofluoresceiny, ≥97%
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
Galactose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
USP
Galaktoza, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
BAPTA-AM, ≥95% (HPLC)
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phospho(enol)pyruvic acid trisodium salt hydrate, ≥97% (enzymatic)
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%