Merck
Glucose transporter and folic acid receptor-mediated Pluronic P105 polymeric micelles loaded with doxorubicin for brain tumor treating.
Journal of drug targeting (2014-05-09)
Jiangxiu Niu, Aidong Wang, Zhongcheng Ke, Zubiao Zheng
ABSTRAKT
In this study, glucose transporter and folic acid (FA) receptor-mediated Pluronic P105 polymeric micelles loaded with DOX (GF-DOX) were prepared for enhancing the blood-brain barrier (BBB) transportation and improving the drug accumulation in the glioma cells. The pH-triggered DOX release of GF-DOX indicating a comparatively fast drug release at weak acidic condition and stable state of the carrier at physiological environment. The transport of GF-DOX across the in vitro BBB model showed that GF-DOX exhibited higher BBB transportation ability with the transporting ratio of 21.47% in 4 h. The carrier was internalized into C6 glioma cells upon crossing the BBB model for the combined effect of the brain targeting by transportation of glucose transporter and active tumor cell targeting by FA receptor-mediated endocytosis. Moreover, minimized weight changes and high suppression ratio of tumor growth were observed after intravenous injection of GF-DOX. In conclusion, the glucose transporter and FA dual-targeting micelles would provide a safe and effective strategy for new modalities to treat brain tumor.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
p-Toluenesulfonyl chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
p-Toluenesulfonyl chloride, reagent grade, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%