Merck
Provirus activation plus CD59 blockage triggers antibody-dependent complement-mediated lysis of latently HIV-1-infected cells.
Journal of immunology (Baltimore, Md. : 1950) (2014-08-26)
Jie Lan, Kai Yang, Daniel Byrd, Ningjie Hu, Tohti Amet, Nicole Shepherd, Mona Desai, Jimin Gao, Samir Gupta, Yongtao Sun, Qigui Yu
ABSTRAKT
Latently HIV-1-infected cells are recognized as the last barrier toward viral eradication and cure. To purge these cells, we combined a provirus stimulant with a blocker of human CD59, a key member of the regulators of complement activation, to trigger Ab-dependent complement-mediated lysis. Provirus stimulants including prostratin and histone deacetylase inhibitors such as romidepsin and suberoylanilide hydroxamic acid activated proviruses in the latently HIV-1-infected T cell line ACH-2 as virion production and viral protein expression on the cell surface were induced. Romidepsin was the most attractive provirus stimulant as it effectively activated proviruses at nanomolar concentrations that can be achieved clinically. Antiretroviral drugs including two protease inhibitors (atazanavir and darunavir) and an RT inhibitor (emtricitabine) did not affect the activity of provirus stimulants in the activation of proviruses. However, saquinavir (a protease inhibitor) markedly suppressed virus production, although it did not affect the percentage of cells expressing viral Env on the cell surface. Provirus-activated ACH-2 cells expressed HIV-1 Env that colocalized with CD59 in lipid rafts on the cell surface, facilitating direct interaction between them. Blockage of CD59 rendered provirus-activated ACH-2 cells and primary human CD4(+) T cells that were latently infected with HIV-1 sensitive to Ab-dependent complement-mediated lysis by anti-HIV-1 polyclonal Abs or plasma from HIV-1-infected patients. Therefore, a combination of provirus stimulants with regulators of complement activation blockers represents a novel approach to eliminate HIV-1.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
SAHA, ≥98% (HPLC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)