Merck
  • Development and validation of a HPLC-MS/MS method for the determination of phytolaccagenin in rat plasma and application to a pharmacokinetic study.
Development and validation of a HPLC-MS/MS method for the determination of phytolaccagenin in rat plasma and application to a pharmacokinetic study.
Journal of pharmaceutical and biomedical analysis (2015-01-13)
Fenghuan Wei, Ravi Shankar Prasad Singh, Matthias Fueth, Steven Swarts, Paul Okunieff, Hartmut Derendorf
ABSTRAKT
Radix Phytolaccae (the dried root of Phytolacca acinosa Roxb. or Phytolacca americana L.) is widely used in East Asian countries for the treatment of inflammation-related diseases. The active component of Radix Phtolaccae is Phytolcaccagenin a triterpenoid saponin. Phytolcaccagenin has anti-inflammatory activities that exceed those of Esculentoside A and its derivatives regarding suppression of LPS-induced inflammation, and has a lower toxicity profile with less hemolysis. To date, no information is available about analytical method and pharmacokinetic studies of phytolaccagenin. To explore PK profile of this compound, a HPLC-MS/MS assay of phytolaccagenin in rat plasma was developed and validated. The method was fully validated according to FDA Guidance for industry. The detection was performed by a triple-quadrupole tandem mass spectrometer with multiple reactions monitoring (MRM) in positive ion mode via electrospray ionization. The monitored transitions were m/z 533.2>515.3 for Phytolcaccagenin, and 491.2>473.2 for I.S. The analysis was performed on a Symmetry C18 column (4.6 mm × 50 mm, 3.5 μm) using gradient elution with the mobile phase consisting of acetonitrile and 0.1% formic acid water at a flow rate of 1 ml/min with a 1:1 splitter ratio. The method was validated with a LLOQ of 20 ng/ml and an ULOQ of 1000 ng/ml. The response versus concentration data were fitted with 1/x weighting and the correlation coefficient (r) were greater than 0.999. The average matrix effect and the average extraction recovery were acceptable. This validation in rat plasma demonstrated that phytolaccagenin was stable for 30 days when stored below -20°C, for 6h at room temperature (RT, 22°C), for 12 h at RT for prepared control samples in auto-sampler vials, and during three successive freeze/thaw cycles results at -20°C. The validated method has been successfully applied to an intravenous bolus pharmacokinetic study of phytolaccagenin in male Sprague-Dawley rats (10 mg/kg, i.v.). Blood samples taken from 0 to 24h after injection were collected, and data analyzed with WinNonlin. The half-life and clearance were 1.4±0.9 h and 2.1±1.1 L/h/kg, respectively.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
USP
Alkohol metylowy, United States Pharmacopeia (USP) Reference Standard