Merck
One-staged aptamer-based isolation and application of endothelial progenitor cells in a porcine myocardial infarction model.
Nucleic acid therapeutics (2014-12-11)
Christoph Haller, Bianka Sobolewska, David Schibilsky, Meltem Avci-Adali, Christian Schlensak, Hans-Peter Wendel, Tobias Walker
ABSTRAKT
A multitude of stem cell types has been extensively studied and used for myocardial regenerative therapy. Amongst these endothelial progenitor cells form a promising source. In our present study, we investigated a one-staged approach for isolation and application of autologous endothelial progenitor cells in a pig model of myocardial infarction. Endothelial progenitor cell isolation was performed by immediately preprocedural bone marrow aspiration and consecutive positive selection by aptamer-based magnetic cell sorting. Animals were divided in three groups receiving endothelial progenitor cells, saline, or no intramyocardial injection respectively. Postprocedural follow-up consisted of weekly echocardiographic evaluations. Postmortem histological analysis after four weeks focused on detection of transplanted PKH26-positive endothelial progenitor cells and neovascularization within the infarcted myocardium. A significant difference in left ventricular ejection fraction could not be shown between the three groups. PKH26-stained endothelial progenitor cells could be found in the endothelial progenitor cells transplanted group, although detection was scarce. Large-sized capillaries were found to be significantly more in endothelial progenitor cells treated myocardium. The one-stage approach of endothelial progenitor cells isolation and application presented herein offers a new therapeutic concept. Even though a beneficial impact on myocardial function could not be assessed, increased neovascularization may indicate positive effects on remodeling processes. Being able to harvest endothelial progenitor cells right before application provides a wider scope of action in urgent cases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Propofol, European Pharmacopoeia (EP) Reference Standard
Supelco
Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Sigma-Aldrich
Potassium chloride, Molecular Biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Supelco
Potassium chloride solution, ~3 M KCl, saturated with silver chloride