Merck
NMDA receptors are upregulated and trafficked to the plasma membrane after sigma-1 receptor activation in the rat hippocampus.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-08-22)
Mohan Pabba, Adrian Y C Wong, Nina Ahlskog, Elitza Hristova, Dante Biscaro, Wissam Nassrallah, Johnny K Ngsee, Melissa Snyder, Jean-Claude Beique, Richard Bergeron
ABSTRAKT
Sigma-1 receptors (σ-1Rs) are endoplasmic reticulum resident chaperone proteins implicated in many physiological and pathological processes in the CNS. A striking feature of σ-1Rs is their ability to interact and modulate a large number of voltage- and ligand-gated ion channels at the plasma membrane. We have reported previously that agonists for σ-1Rs potentiate NMDA receptor (NMDAR) currents, although the mechanism by which this occurs is still unclear. In this study, we show that in vivo administration of the selective σ-1R agonists (+)-SKF 10,047 [2S-(2α,6α,11R*]-1,2,3,4,5,6-hexahydro-6,11-dimethyl-3-(2-propenyl)-2,6-methano-3-benzazocin-8-ol hydrochloride (N-allylnormetazocine) hydrochloride], PRE-084 (2-morpholin-4-ylethyl 1-phenylcyclohexane-1-carboxylate hydrochloride), and (+)-pentazocine increases the expression of GluN2A and GluN2B subunits, as well as postsynaptic density protein 95 in the rat hippocampus. We also demonstrate that σ-1R activation leads to an increased interaction between GluN2 subunits and σ-1Rs and mediates trafficking of NMDARs to the cell surface. These results suggest that σ-1R may play an important role in NMDAR-mediated functions, such as learning and memory. It also opens new avenues for additional studies into a multitude of pathological conditions in which NMDARs are involved, including schizophrenia, dementia, and stroke.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Glycine, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)