Merck
MicroRNA-494 inhibits the growth and angiogenesis-regulating potential of mesenchymal stem cells.
FEBS letters (2015-02-11)
Shiwen Chen, Guangfeng Zhao, Huishuang Miao, Ruijing Tang, Yuxian Song, Yali Hu, Zhiqun Wang, Yayi Hou
ABSTRAKT
Mesenchymal stem cells (MSCs) play an important role in the pathology of preeclampsia (PE). Our previous microarray analysis found that microRNA-494 (miR-494) is highly expressed in decidua-derived MSCs (dMSCs) from PE. We hypothesized that aberrant expression of miR-494 in dMSCs is involved in PE development. In the present study, we found that miR-494 arrests G1/S transition in dMSCs by targeting CDK6 and CCND1. We also found that supernatant from miR-494-overexpressing dMSCs reduces HTR-8/SVneo migration and impairs HUVEC capillary formation by suppressing VEGF. Taken together, we report an unrecognized mechanism of miR-494 affecting dMSC proliferation and function in the pathology of PE.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard