Merck
A yeast-based assay identifies drugs that interfere with immune evasion of the Epstein-Barr virus.
Disease models & mechanisms (2014-02-22)
Cécile Voisset, Chrysoula Daskalogianni, Marie-Astrid Contesse, Anne Mazars, Hratch Arbach, Marie Le Cann, Flavie Soubigou, Sébastien Apcher, Robin Fåhraeus, Marc Blondel
ABSTRAKT
Epstein-Barr virus (EBV) is tightly associated with certain human cancers, but there is as yet no specific treatment against EBV-related diseases. The EBV-encoded EBNA1 protein is essential to maintain viral episomes and for viral persistence. As such, EBNA1 is expressed in all EBV-infected cells, and is highly antigenic. All infected individuals, including individuals with cancer, have CD8(+) T cells directed towards EBNA1 epitopes, yet the immune system fails to detect and destroy cells harboring the virus. EBV immune evasion depends on the capacity of the Gly-Ala repeat (GAr) domain of EBNA1 to inhibit the translation of its own mRNA in cis, thereby limiting the production of EBNA1-derived antigenic peptides presented by the major histocompatibility complex (MHC) class I pathway. Here we establish a yeast-based assay for monitoring GAr-dependent inhibition of translation. Using this assay we identify doxorubicin (DXR) as a compound that specifically interferes with the GAr effect on translation in yeast. DXR targets the topoisomerase-II-DNA complexes and thereby causes genomic damage. We show, however, that the genotoxic effect of DXR and various analogs thereof is uncoupled from the effect on GAr-mediated translation control. This is further supported by the observation that etoposide and teniposide, representing another class of topoisomerase-II-DNA targeting drugs, have no effect on GAr-mediated translation control. DXR and active analogs stimulate, in a GAr-dependent manner, EBNA1 expression in mammalian cells and overcome GAr-dependent restriction of MHC class I antigen presentation. These results validate our approach as an effective high-throughput screening assay to identify drugs that interfere with EBV immune evasion and, thus, constitute candidates for treating EBV-related diseases, in particular EBV-associated cancers.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Methylene blue, certified by the BSC
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride, ≥98%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Idarubicin hydrochloride, solid
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Etoposide, European Pharmacopoeia (EP) Reference Standard
Informacje o cenach i dostępności nie są obecnie dostępne.