Merck
Multimeric grain-marked micelles for highly efficient photodynamic therapy and magnetic resonance imaging of tumors.
International journal of pharmaceutics (2014-05-27)
Ung Yeol Lee, Young Taik Oh, Dongin Kim, Eun Seong Lee
ABSTRAKT
Multimeric grain-marked micelles consisting of an inner core micelle (for Fe3O4 encapsulation) and outer multi-grain micelles (for chlorin e6 (Ce6, a model drug) encapsulation) were fabricated using a micelle-to-micelle conjugation method. Grain micelles (mono-thiol functionalized micelles) were chemically linked to the surface of the core micelle (multi-maleimide functionalized micelle). These micelles enable discrete compartments for Ce6 and iron oxide (Fe3O4) that enable a significantly increased in vivo photodynamic tumor inhibition while preserving high contrast magnetic resonance (MR) imaging of the tumor in vivo.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DL-Dithiothreitol, BioUltra, Molecular Biology, ≥99.5% (RT)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
DL-Dithiothreitol, Molecular Biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
DL-Dithiothreitol, suitable for electrophoresis, ≥99%
Sigma-Aldrich
DL-Dithiothreitol, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0% (RT)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Supelco
Triethylamine, analytical standard
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture