Merck
Hydrogen sulfide delays nicotinamide-induced premature senescence via upregulation of SIRT1 in human umbilical vein endothelial cells.
Molecular and cellular biochemistry (2014-04-15)
Meihua Zheng, Weili Qiao, Jie Cui, Lei Liu, Hong Liu, Zhirong Wang, Changdong Yan
ABSTRAKT
The present study was designed to investigate the effect of hydrogen sulfide on cellular senescence of human umbilical vascular endothelial cells (HUVECs CC-2517) and its underlying mechanism. The premature senescence-like phenotype HUVECs (the fourth passage) was induced by treatment with nicotinamide (NAM, an inhibitor of SIRT1, 5 mmol/L, 12 h). Cells were cultured with sodium hydrosulfide (NaHS, 12.5, 25, 50 and 100 μmol/L) for 48 h in premature senescence-like phenotype HUVECs. The fourth passage of HUVECs was considered as young group. Senescence-associated (SA)-β-galactosidase activities were detected to evaluate cell senescence, and the expression of SA heterochromatin foci (SAHF) was visualized by DAPI DNA staining. The mRNA and protein levels of SIRT1 were detected using RT-PCR and western blotting analysis, respectively. The results showed that β-galactosidase positive cells and the formation of SAHF were markedly increased after treatment with NAM (5 mmol/L) for 12 h. We also found that NaHS (12.5 μmol/L) had no effect on the percentage of SA β-gal positive cells and the expression of SAHF, and the hallmarks decreased at the concentration of 25 and 50 μmol/L, reaching the minimum at 50 μmol/L, while the percentage of SA β-gal positive cells and the expression of SAHF increased at the concentration of 100 μmol/L. Furthermore, we found that both on protein and mRNA levels of SIRT1 in the Y+N+S50 group was significantly increased compared with that in Y+N group. In conclusion, NaHS delays senescence of HUVECs induced by NAM via upregulation of SIRT1 expression.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Resveratrol, analytical standard
Supelco
Resveratrol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Niacynamid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Resveratrol, European Pharmacopoeia (EP) Reference Standard