Merck
  • Rapid and sensitive determination of diacetylpolyamines in human fingernail by ultraperformance liquid chromatography coupled with electrospray ionization tandem mass spectrometry.
Rapid and sensitive determination of diacetylpolyamines in human fingernail by ultraperformance liquid chromatography coupled with electrospray ionization tandem mass spectrometry.
European journal of mass spectrometry (Chichester, England) (2014-01-01)
Jun Zhe Min, Yuka Morota, Ying-Zi Jiang, Gao Li, Dongzhou Kang, Hai-fu Yu, Koichi Inoue, Kenichiro Todoroki, Toshimasa Toyo'oka
ABSTRAKT
A rapid and sensitive ultraperformance liquid chromatography coupled with electrospray ionization tandem mass spectrometry (UPLC-ESI-MS/MS) method has been developed and validated for quantitatively determining diacetylpolyamines in the human fingernail. N(1),N(8)-diacetylspermidine (DiAct-Spd), N(1),N(12)- diacetylspermine (DiAct-Spm) and 1,6-diaminohexane (DAH) the [internal standard (IS)] were extracted from human fingernail samples by MeOH: 5 M HCl solution, followed by 4-(N,N-dimethylaminosulfonyl)-7-fluoro- 2,1,3-benzoxadiazole (DBD-F) derivatization, and then separated on an ACQUITY BEH C18 column with a gradient elution of acetonitrile and water containing 0.1% formic acid. The derivatives of the diacetylpolyamines were fully separated within a short run time (3.0 min). The triple quadrupole mass spectrometric detection was performed in the multiple reactions monitoring (MRM) mode by the UPLC-ESI- MS/MS system in the positive ionization mode. MRM using the fragmentation transitions of m/z 455.20→ 100.07, 737.25 → 100.07 and 567.10 → 479.07 in the positive ESI mode was performed to quantify DiAct-Spd, DiAct-Spm and IS, respectively. The calibration curve is between 0.04 ng mL(-1) for DiAct-Spd and DiAct-Spm. The detection limits (signal to noise ratio of five) were 5-10 pg mL(-1). A good linearity was achieved from the calibration curves (r(2) >0.9999), and the intra-day and inter-day assay precisions were less than 7.06%. Furthermore, the recoveries (%) of the diacetylpolyamines spiked in the human fingernails were 79.18-97.11. The present method proved that the high sensitivity is characterized by the specificity and feasibility of the sample analysis. Consequently, the proposed method was used to analyze human fingernail samples from 15 lung- cancer patients and 22 healthy volunteers. Diacetylpolyamines were detected from the fingernails of the lung- cancer patients for the first time. The concentration of DiAct-Spd in the lung-cancer patient group tended to be higher than those in the healthy volunteers.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology