Merck
Formulation and pharmacokinetics of thermosensitive stealth® liposomes encapsulating 5-Fluorouracil.
Pharmaceutical research (2014-11-25)
Chantal Al Sabbagh, Nicolas Tsapis, Anthony Novell, Patricia Calleja-Gonzalez, Jean-Michel Escoffre, Ayache Bouakaz, Hélène Chacun, Stéphanie Denis, Juliette Vergnaud, Claire Gueutin, Elias Fattal
ABSTRAKT
We optimize the encapsulation and investigate the pharmacokinetics of 5-Fluorouracil (5-FU) delivered by thermosensitive stealth(®) liposomes (TSLs) designed to trigger drug release upon hyperthermia using focused ultrasound (FUS). 5-FU was encapsulated into liposomes made of 1,2-Dipalmitoyl-sn-glycero-3-phosphocholine/cholesterol/1,2-Distearoyl-sn-glycero-3-phosphoethanolamine-N-PEG2000 either as a free molecule or complexed with copper-polyethylenimine. Heat-triggered drug release was evaluated using either a water bath or FUS. Formulation cytotoxicity was assessed on HT-29 cell line by MTS assay. Pharmacokinetics and biodistribution of 5-FU were evaluated in HT-29-tumor bearing mice. 5-FU was easily encapsulated using the lipid hydration method (encapsulation efficacy of 13%) but poorly retained upon dilution. 5-FU complexation with copper-polyethylenimine improved 5-FU retention into liposomes and allowed to obtain an encapsulation efficacy of 37%. At 42°C, heat-triggered 5-FU release from TSLs was 63% using a water bath and 68% using FUS, within 10 min, whereas it remained below 20% for the non-thermosensitive formulation. The MTS assay revealed that formulation toxicity arose from 5-FU and not from the excipients. In addition, 5-FU complex encapsulation into TSLs induces a reduction of the IC50 from 115 down to 49 μM. Pharmacokinetics reveals a longer circulation of encapsulated 5-FU and a more important body exposure, although tumor passive targeting is not significantly higher than free 5-FU. Complexation of 5-FU with copper-polyethylenimine appears an interesting strategy to improve 5-FU retention into TSLs in vitro and in vivo. TSLs allow heat-triggered release of the drug within 10 min at 42°C, a reasonable time for future in vivo experiments.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
USP
Fluorouracyl, United States Pharmacopeia (USP) Reference Standard
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Supelco
5-Fluorouracil, analytical standard
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
Supelco
D-(+)-Glucose, analytical standard
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%