Merck
pH-responsive, lysine-based hydrogels for the oral delivery of a wide size range of molecules.
International journal of pharmaceutics (2014-12-10)
Kira A Watkins, Rongjun Chen
ABSTRAKT
Hydrogels synthesized from poly(l-lysine isophthalamide) (PLP) crosslinked with l-lysine methyl ester were investigated as drug delivery systems for a wide size range of molecules (0.3-2000kDa). PLP is an anionic, pseudo-peptidic polymer that is an ideal hydrogel backbone due to its pH-responsiveness and amphiphilicity. Drug loading and release were evaluated for various model drugs: hydrophobic fluorescein (Mw=332Da) and hydrophilic fluorescein isothiocyanate-dextran (FITC-Dex Mw=10kDa, 150kDa, 500kDa, and 2000kDa). Weight incorporation was high, up to 22.8±3.1%. Release after 24h in pH 7.4 was in the range from 70.4±1.2% to 91.6±0.8% for all model drugs. In contrast, drug release after 24h in pH 3.0 was significantly lower, less than 8% for fluorescein, 500kDa, and 2000kDa FITC-Dex. Thus, the adaptability of these novel hydrogels to both hydrophobic and hydrophilic molecules, spanning a wide size range, suggests their use as a platform delivery system. This is also the first known hydrogel system for the oral delivery of payloads larger than 70kDa, which, combined with triggered release in response to pH changes along the gastrointestinal tract, indicates that these hydrogels have promising applications in oral drug delivery.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Isophthaloyl chloride, ≥99%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Trisodium citrate dihydrate, meets USP testing specifications
Sigma-Aldrich
L-Lysine methyl ester dihydrochloride
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, ≥98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether