Merck
Development of a capillary electrophoretic method for determination of plasma clearance of iohexol in dogs and cats.
Biomedical chromatography : BMC (2014-08-12)
Alexander Hellqvist, Reidun Heiene, Siegrid De Baere, Siska Croubels, Ylva Hedeland
ABSTRAKT
Renal function can be monitored by estimation of the glomerular filtration rate (GFR), for example, through measurement of the plasma clearance of a marker that is freely filtrated through the kidney without reabsorption. It has been proposed that iohexol is the most accurate marker for GFR determination in cats and dogs. However, there is a need for a validated capillary electrophoretic method that covers the concentration range for a full curve clearance estimate of iohexol. In the final method, the plasma samples were protein precipitated and the supernatant was analyzed in a background electrolyte containing borate buffer (0.06 m, pH 10.0). The method developed was proved to be linear (concentration range 18- 2900 mg/L) and had a good precision (e.g. 2.3-2.9% at 88 mg/L) and accuracy (e.g. 101-105% at 88 mg/L). Finally, the method was compared with a previously published and validated HPLC-UV method by parallel analysis of clinical plasma samples from dogs and cats administered Omnipaque®. This comparison showed excellent agreement between the two methods and no proportional or systematic error was observed. The proposed method is simple and has a low cost per sample, which makes it applicable for routine analysis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Supelco
Benalaxyl-M, PESTANAL®, analytical standard
Sigma-Aldrich
Boric acid, BioUltra, Molecular Biology, ≥99.5% (T)
Supelco
Acrylamide solution, 40% in H2O
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Sigma-Aldrich
Boric acid, BioReagent, Molecular Biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Boric acid, tablet, 1 g boric acid per tablet
Sigma-Aldrich
Boric acid, ReagentPlus®, ≥99.5%
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Boric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Boric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, ≥99.8%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
USP
Alkohol metylowy, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)