Merck
  • Polyelectrolyte complexes of poly[(2-dimethylamino) ethyl methacrylate]/chondroitin sulfate obtained at different pHs: I. Preparation, characterization, cytotoxicity and controlled release of chondroitin sulfate.
Polyelectrolyte complexes of poly[(2-dimethylamino) ethyl methacrylate]/chondroitin sulfate obtained at different pHs: I. Preparation, characterization, cytotoxicity and controlled release of chondroitin sulfate.
International journal of pharmaceutics (2014-10-15)
Letícia C Bonkovoski, Alessandro F Martins, Ismael C Bellettini, Francielle P Garcia, Celso V Nakamura, Adley F Rubira, Edvani C Muniz
ABSTRAKT
For the first time, polyelectrolyte complex based on poly[(2-dimethylamino) ethyl methacrylate] (PDMAEMA) and chondroitin sulfate (CS) was prepared. The properties of novel material and precursors were investigated by WAXS, FTIR, TGA, SEM and DLS analysis. The PDMAEMA/CS PECs presented hydrophilic-hydrophobic transition at pHs 6.0, 7.0 and 8.0 whereas the non-complexed PDMAEMA showed such a transition at pH 8.0 and not at pHs 6.0 and 7.0. Studies of CS release from PECs at pHs 6 and 8 confirmed that the samples possess the potential to release the CS in alkaline and not in acidic conditions. Since PECs are thermo-responsive due to the reduction of LCST caused by the increase in pH, the release of CS was dependent on temperature and pH factors. Cytotoxicity assays using healthy VERO cells showed that the complexation between CS and PDMAEMA increased the PECs' biocompatibility related to PDMAEMA. However, the biocompatibility depends on the amount of CS present in the PECs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
SAFC
Glycine
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Methylene blue, certified by the BSC
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade
2-(Dimethylamino)ethyl methacrylate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution