Merck
Effects of intestinal microbiota on the bioavailability of geniposide in rats.
Journal of agricultural and food chemistry (2014-09-27)
Ming Ji Jin, In Sook Kim, Dong-Hyun Kim, Hye Hyun Yoo
ABSTRAKT
This study investigated the effects of intestinal microbiota on the metabolism of geniposide by using a rat model treated with a mixture of antibiotics. The plasma concentration of geniposide was determined after oral administration in control and antibiotics-treated rats by using liquid chromatography-tandem mass spectrometry. The maximum plasma concentrations (Cmax) of geniposide in control and antibiotics-treated rats were 0.91 ± 0.26 and 1.01 ± 0.04 μg/mL, respectively, and the area under the curve (AUC) values were 7.34 ± 3.32 and 11.9 ± 2.1 μg·h/mL (p < 0.05), respectively. The levels of geniposide in rat feces were 0.64 and 15.6 mg, respectively, in the control and antibiotics-treated groups. Thus, the systemic exposure of geniposide was greater in the antibiotics-treated rats. This may be due to the antibiotic-induced suppression of the metabolic activities of the intestinal microbiota. These results suggest that the gut microbiota may have an impact on the bioavailability of geniposide.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
USP
Alkohol metylowy, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Digoxin for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Supelco
Digoxin, analytical standard
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material