Merck
Contribution of glucose to crystallization of phenytoin in injectable dosage form by dilution with infusion fluids.
Chemical & pharmaceutical bulletin (2014-10-03)
Yoshinori Onuki, Naoki Hasegawa, Mayumi Ikegami-Kawai, Takako Suematsu, Satoshi Sakurai, Shunsuke Shirozu, Masayoshi Tsubuki, Yasuko Obata, Kozo Takayama
ABSTRAKT
The crystallization of phenytoin occurring after its dilution with infusion fluid is a major concern in the clinical use of injectable phenytoin. To gain further understanding of the crystallization, this study assessed details of the involvement of glucose in this action. For sample preparation, phenytoin crystals were created by diluting the injectable phenytoin with infusion fluids with different glucose concentrations at different temperature, and then the characteristics of the crystallization (e.g., crystal size in the long direction, accumulated amount over 24 h, and crystallization rate constant) were measured. Results of the analysis of variance indicated that the glucose concentration and temperature had significant impacts on the crystallization. The mode of action of the glucose concentration was suggested to be different from that of the incubation temperature. This study also examined the molecular mobility of components (i.e., glucose, propylene glycol, phenytoin) in the admixtures using diffusion NMR techniques. The findings will provide valuable information for the clinical use of injectable phenytoin.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
USP
Glikol propylenowy, United States Pharmacopeia (USP) Reference Standard
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC