Merck
Improved assays for xenosensor activation based on reverse transfection.
Toxicology in vitro : an international journal published in association with BIBRA (2015-07-19)
Jenni Küblbeck, Teemu Anttila, Juha T Pulkkinen, Paavo Honkakoski
ABSTRAKT
Discovery of receptor-dependent mechanisms for regulation of drug metabolism has provided a new way to evaluate the propensity of drug candidates to cause induction of cytochrome P450 enzymes. Therefore, receptor-based reporter assays have become common in early stages of drug development projects and in mechanistic studies. Here, we report a reverse transfection system to conduct activation assays for human xenosensors AhR, CAR and PXR. The assay format is based on long-term stability and uniformity of DNA/carrier complexes on culture plates, avoiding multiple stages and variation inherent in conventional transfection methods. Consequently, these improved assays are streamlined, reproducible and formally validated with Z' factors exceeding 0.5. This novel reverse transfection system is expected to find use in diverse areas of early drug development such prediction of CYP induction, evaluation of species differences and in mechanistic studies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
17α-Ethynylestradiol, ≥98%
Sigma-Aldrich
β-Naphthoflavone, ≥98%
Sigma-Aldrich
Diethylstilbestrol, ≥99% (HPLC)
Sigma-Aldrich
Clotrimazole
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Mifepristone, ≥98%
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Hyperforin, ≥85% (HPLC)