Merck
Relation between Ku80 and microRNA-99a expression and late rectal bleeding after radiotherapy for prostate cancer.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology (2015-05-06)
Masanori Someya, Hiroyuki Yamamoto, Masanori Nojima, Masakazu Hori, Kunihiko Tateoka, Kensei Nakata, Masaru Takagi, Masato Saito, Naoki Hirokawa, Takashi Tokino, Koh-Ichi Sakata
ABSTRAKT
Late rectal bleeding is one of the severe adverse events after radiotherapy for prostate cancer. New biomarkers are needed to allow a personalized treatment. Four patients each with grade 0-1 or grade 2-3 rectal bleeding were randomly selected for miRNA array to examine miRNA expression in peripheral blood lymphocytes (PBLs). Based on results of miRNA array, 1 of 348 miRNAs was selected for microRNA assays. Then, expression of DNA-dependent protein kinase mRNA and miR-99a was analyzed in the PBLs of 97 patients. PBLs were exposed to 4Gy of X-ray ex-vivo. In the discovery cohort, grade 2-3 rectal bleeding was significantly higher in the Ku80 <1.09 expression group compared with ⩾1.09 group (P=0.011). In radiation-induced expression of miR-99a, grade 2-3 rectal bleeding was significantly higher in the miR-99a IR(+)/IR(-) >0.93 group compared with ⩽0.93 group (P=0.013). Most patients with grade 2-3 rectal bleeding were in the group with low Ku80 and high miR-99a expression. In the validation cohort, similar results were obtained. A combination of low Ku80 expression and highly-induced miR-99a expression could be a promising marker for predicting rectal bleeding after radiotherapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O