Merck
  • Antioxidant potential, in vitro cytotoxicity and apoptotic effect induced by crude organic extract of Anthracophyllum lateritium against RD sarcoma cells.
Antioxidant potential, in vitro cytotoxicity and apoptotic effect induced by crude organic extract of Anthracophyllum lateritium against RD sarcoma cells.
BMC complementary and alternative medicine (2015-11-08)
Dilusha M Fernando, Ravi L C Wijesundera, Preethi Soysa, Dilip de Silva, Chandrika M Nanayakkara
ABSTRAKT
Macrofungi have an established history of use in traditional oriental medicine. Anthracophyllum lateritium is a terrestrial macrofungus found in the dry zone forest reserves in Sri Lanka. Yet there are no scientific reports on bioactive properties of this species. Hence, the current study was aimed at determining the antioxidant potential, in vitro antiproliferative activity and apoptotic effect induced by crude methanolic extract of A. lateritium against RD sarcoma cell line. The crude extract of A. lateritium was dissolved in methanol (MEFCA) and antioxidant activity was evaluated using in vitro assays: inhibition of DPPH (1,1-diphenyl-2-picrylhydrazyl) radical scavenging, ferric ion reducing power and 2-deoxy-D-ribose degradation assay. Total phenol and flavonoid contents of MEFCA were assayed using folin Ciocalteu method and aluminium chloride colorimetric method. In vitro cytotoxicity was determined using MTT assay against RD cells after 24 h exposure to MEFCA. Ethidium bromide/ acridine orange staining, DNA fragmentation and protein synthesis experiments were used to study the apoptotic features and antiproliferative activities of the treated cells. Glutathione assay and griess nitrite assay were used to analyze the reduced glutathione content and liberation of nitric oxide from apoptotic cells. MEFCA showed promising antioxidant activity with EC50 values of 8.00 ± 0.35 μg/mL for DPPH scavenging and 83.33 ± 0.45 μg/mL for 2-deoxy-D-ribose degradation assay. The phenolic content was 265.15 ± 0.46 of (w/w) % of Gallic acid equivalents and flavonoid content was 173.01 ± 0.35 of (w/w) % of Epigallocatechingallate. A. lateritium showed strong in vitro cytotoxic activity with an EC50 of 18.80 ± 4.83 μg/mL for MTT assay against RD cells. Ethidium bromide/acridine orange staining and DNA fragmentation indicated the apoptotic features of treated cells. Protein levels showed a dose dependent decrease supporting the fact that A. lateritium induces apoptosis of treated cells. Glutathione content and nitric oxide content of cells exhibited a dose dependent increase suggesting the apoptosis of RD cells was mediated by both nitrie ions and nitric oxide. The crude extract of the A. lateritium exhibited potent antioxidant, antiproliferative activity and apoptotic effect against RD cells providing supportive evidence for the ethnopharmacological use of this fungus in control of oxidative damage and remedy of cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Aluminum chloride, ReagentPlus®, 99%