Merck
Effects of oxygen on the antigenic landscape of prostate cancer cells.
BMC research notes (2015-11-20)
Tangeng Ma, Claire A Schreiber, Gaylord J Knutson, Abdelouahid El Khattouti, Marcelo J Sakiyama, Mohamed Hassan, Mary Christine Charlesworth, Benjamin J Madden, Xinchun Zhou, Stanimir Vuk-Pavlović, Christian R Gomez
ABSTRAKT
Use of allogeneic cancer cells-based immunotherapy for treatment of established prostate cancer (PCa) has only been marginally effective. One reason for failure could stem from the mismatch of antigenic signatures of vaccine cells and cancer in situ. Hence, it is possible that vaccine cells expressed antigens differently than tumor cells in situ. We hypothesized that cells grown in vitro at low oxygen tension (pO2) provide a better antigen match to tumors in situ and could reveal a more relevant antigenic landscape than cells grown in atmospheric pO2. We tested this hypothesis by comparing PCa cells propagated at pO2 = 2 kPa and 20 kPa. To identify potential tumor-associated antigens (TAAs), we prepared PCa cell lysates, resolved them by two-dimensional electrophoresis and immunoblotting using spontaneous antibodies from plasma derived from PCa patients and control subjects. Antibody-labeled spots were analyzed by MALDI-TOF mass spectrometry and validated by ELISA. We selected hypoxia-regulated HSP70 and hnRNP L and hypoxia-independent HSP60 and determined the frequency of plasma samples reacting with these molecules. Frequency of HSP60-reactive plasma was low in healthy controls [1.3 % (1/76)], while it was elevated in PCa patients [13.0 % (7/54); p < 0.05]. These data suggest a humoral immune response to HSP60 in PCa. Levels of autoantibodies to HSP70 did not differ from healthy controls [3.7 % (2/54)] in PCa patients [5.3 % (2/38)]. Similarly, hnRNP L autoantibodies did no differ between healthy controls [6.1 % (3/49)] and PCa patients [5.3 % (2/38)]. Overall our results suggest the value of hypoxia as a modifier of the cellular and antigenic landscape of PCa cells. By modifying the immune reactivity of PCa cells in culture, manipulation of pO2 can be proposed as a new avenue for improving diagnosis, prognosis and immunotherapy for PCa.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)