Merck
Comparative proteomic analysis provides insight into cadmium stress responses in brown algae Sargassum fusiforme.
Aquatic toxicology (Amsterdam, Netherlands) (2015-04-02)
Aiqin Zhang, Tao Xu, Huixi Zou, Qiuying Pang
ABSTRAKT
Sargassum fusiforme is one of the most widely consumed seaweeds in China, Korea and Japan. In this work, we performed growth analysis and comparative proteomics to investigate the molecular mechanisms of the response to 1 day and 5 days Cd stress in S. fusiforme. Our results showed a significant decrease in growth rate and an increase in Cd ion content in S. fusiforme in response to Cd treatment. Comparative proteomic analysis revealed 25 and 51 differentially expressed protein spots in S. fusiforme under 1 day and 5 days Cd stress, respectively. A great number of these proteins was metabolic enzymes involved in carbohydrate metabolism and energy metabolism. Many proteins involved in the processing of genetic information showed a decrease in abundance under 1 day Cd stress. In contrast, 9 of the identified protein spots primarily involved in genetic information processing and carbohydrate metabolism were greatly enriched under 5 days Cd stress. Overall, our investigation indicated that Cd stress negatively affects the metabolic activity of S. fusiforme through the down-regulation of key metabolic enzymes. In addition, S. fusiforme may adapt to 5 days Cd stress by promoting consumption of photoassimilates through the up-regulation of glycolysis and the citrate cycle to supply energy for survival.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O