Merck
Cantharidin modulates the E2F1/MCM7-miR-106b-93/p21-PTEN signaling axis in MCF-7 breast cancer cells.
Oncology letters (2016-01-02)
Hui Zhang, Xiuli Yan
ABSTRAKT
Cantharidin (CTD) is one of numerous natural products used in traditional Chinese medicine for the treatment of cancer. The aim of the present study was to investigate the effects of CTD on changes in the expression of microRNAs (miRNAs/miRs) and to explore its anti-proliferative effect on MCF-7 breast cancer cells. The proliferation of MCF-7 cells was measured by performing an MTT assay. MCF-7 cells were treated with various concentrations of CTD for 48 h, and the expression profiles of miRNAs in CTD-treated and -untreated MCF-7 cells were detected using miRNA microarray chips. The array data were confirmed by reverse transcription-quantitative polymerase chain reaction and protein expression levels were measured by western blot analysis. The 50% inhibitory concentration of CTD was 1.75 µg/ml following treatment for 48 h and CTD significantly inhibited the proliferation of MCF-7 cells in a dose-dependent manner (P<0.01). Furthermore, microarray analysis identified 35 miRNAs that were up-regulated (fold change ≥2.0 and P<0.01) and 45 miRNAs that were down-regulated (fold change ≤ 0.5 and P<0.01) in response to CTD treatment. Thus, numerous CTD-induced miRNAs appeared to be associated with breast cancer. Notably, CTD repressed the expression of miR-106b-93, its host gene MCM7 and its transcription factor E2F1. In addition, CTD induced an increase in the protein expression levels of miR-106b-93 target genes p21 and phosphatase and tensin homolog. These observations suggested that the modulation of miRNA expression is an important mechanism underlying the biological effects of CTD in breast cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
SAFC
L-Glutamine
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®