Merck
Cross-Reactivity Between Influenza Matrix- and HIV-1 P17-Specific CTL-A Large Cohort Study.
Journal of acquired immune deficiency syndromes (1999) (2015-04-23)
Angela G Hückelhoven, Jennifer K Etschel, Silke Bergmann, Kathrin Zitzelsberger, Sandra M Mueller-Schmucker, Ellen G Harrer, Thomas Harrer
ABSTRAKT
It has been reported that HIV-1-specific cytotoxic T cells (CTL) recognizing the HLA-A2-restricted p17 epitope SLYNTVATL (SL9) can cross-react with the HLA-A2-restricted influenza matrix epitope GILGFVFTL (GL9). So far, the prevalence of GL9-cross-reacting HIV-1-specific CTL in larger cohorts of HIV-1-infected patients is unknown, and there are no data yet on whether SL9/GL9-cross-reactive CTL may influence the course of HIV-1 infection. We analyzed the presence of SL9/GL9-cross-reacting CTL in a cohort of 175 HLA-A2-positive HIV-1-infected patients. Peripheral blood mononuclear cells were stimulated in vitro with SL9 and GL9 peptides, and outgrowing cell lines regarding cross-reactivity and recognition of viral variants in γ-interferon enzyme-linked immunospot assays were analyzed. SL9- and GL9-specific CTL could be generated in 52.6% and 53.7% of 175 patients, respectively. Both SL9- and GL9-specific CTL were more frequently observed in patients on antiretroviral therapy (ART). Of the 92 SL9-specific CTL and the 94 GL9-specific CTL, 65.2% and 66%, respectively, showed at least partial SL9/GL9 cross-reactivity. SL9/GL9-cross-reactive CTL could be detected in 42.9% of the 175 patients. Recognition of SL9 was associated with lower viral loads and higher CD4 cell counts in patients on ART. Patients with GL9/SL9 cross-reactivity displayed similar CD4 cell counts than patients without GL9/SL9-cross-reactive cells. GL9/SL9-cross-reactive cells were associated with higher viral loads in patients on ART. Partially SL9/GL9-cross-reactive CTL are frequently observed in HIV-1-infected patients. So far, we could not detect a significant influence of the presence of SL9/GL9-cross-reacting CTL on the course of HIV-1 infection.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, carrier free
Sigma-Aldrich
IL-2 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
SAFC
L-Glutamine
Sigma-Aldrich
Interleukin-2, human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O