Merck
  • Effect of refrigerated storage on probiotic viability and the production and stability of antimutagenic and antioxidant peptides in yogurt supplemented with pineapple peel.
Effect of refrigerated storage on probiotic viability and the production and stability of antimutagenic and antioxidant peptides in yogurt supplemented with pineapple peel.
Journal of dairy science (2015-07-06)
B N P Sah, T Vasiljevic, S McKechnie, O N Donkor
ABSTRAKT
Fruit by-products are good resources of carbohydrates, proteins, vitamins, and minerals, which may function as growth nutrients for probiotic bacteria. This research aimed at evaluating effects of pineapple peel powder addition on the viability and activity of Lactobacillus acidophilus (ATCC 4356), Lactobacillus casei (ATCC393), and Lactobacillus paracasei ssp. paracasei (ATCC BAA52) in yogurts throughout storage at 4°C for 28d. Plain and probiotic yogurts supplemented with or without pineapple peel powder or inulin were prepared. The probiotic counts in supplemented yogurts at 28d of storage ranged from 7.68 and 8.03 log cfu/g, one log cycle higher compared with nonsupplemented control yogurt. Degree of proteolysis in synbiotic yogurts was significantly higher than plain yogurts and increased substantially during storage. Crude water-soluble peptide extract of the probiotic yogurt with peel possessed stronger antimutagenic and antioxidant activities [evaluated measuring reducing power and scavenging capacity of 1,1-diphenyl-2-picrylhydrazyl; 2,2'-azino-bis(3-ethyl benzothiazoline-6-sulfonic acid), and hydroxyl radicals] than control and maintained during storage. Pineapple peel, a by-product of juice production, could be proposed as a prebiotic ingredient in the manufacture of yogurts with enhanced nutrition, and functionality.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Octylphosphonic acid, 97%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Phthaldialdehyde, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)