Merck
A new biodegradable and biocompatible gadolinium (III) -polymer for liver magnetic resonance imaging contrast agent.
Magnetic resonance imaging (2015-04-04)
Yan Xiao, Rong Xue, Tianyan You, Xiaojing Li, Fengkui Pei
ABSTRAKT
A new biodegradable and biocompatible gadolinium (III) -copolymer (ACL-A2-DOTA-Gd) has been developed as a potential liver magnetic resonance imaging (MRI) contrast agent. ACL-A2-DOTA-Gd consisted of a poly (aspartic acid-co-leucine) unit bound with 1,4,7,10-tetraazacyclododecan-1,4,7,10-tetraacetic acid-gadolinium (Gd-DOTA) via the linkage of ethylenediamine. In vitro, the biodegradable experiment and cytotoxicity assay showed the biodegradability and biocompatibility of this gadolinium-polymer. ACL-A2-DOTA-Gd presented an increase in relaxivity of 2.4 times than the clinical Gd-DOTA. In vivo, gadolinium (III)-copolymer was mainly accumulated in the liver, and it could be excreted via the renal and hepatobiliary mechanism. The average enhancement of ACL-A2-DOTA-Gd (60.71±5.93%, 50-80 min) in liver was 2.62-fold greater than that of Gd-DOTA (23.16±3.55%, 10-30 min). ACL-A2-DOTA-Gd could be as a potential liver MRI contrast agent with a long time-window.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)
SAFC
L-Aspartic acid
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid, ≥97.0% (CHN)
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Gadolinium(III) chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Gadolinium(III) chloride solution, suitable for NMR (reference standard), 0.1 mg/mL in D2O (99.9 atom % D), Methanol-13C 0.1 % (99 atom % 13C), water 1 %, NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Gadolinium(III) chloride solution, suitable for NMR (reference standard), 0.1 mg/mL in D2O (99.9 atom % D), water 0.1 %, NMR tube size 6.5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Ethylenediamine, BioXtra
Sigma-Aldrich
Ethylenediamine, meets USP testing specifications
Sigma-Aldrich
Ethylenediamine solution, technical, 75-80%
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Gadolinium, chips