Merck
Cyclic stretch enhances bone morphogenetic protein-2-induced osteoblastic differentiation through the inhibition of Hey1.
International journal of molecular medicine (2015-12-10)
Zhaobin Zeng, Xiao Yin, Xiaodong Zhang, Da Jing, Xue Feng
ABSTRAKT
Substantial evidence has indicated that osteoblastic differentiation may be regulated by mechanical loads or bone morphogenetic protein-2 (BMP-2). BMP-2-induced in vivo osteogenesis can be significantly enhanced in the presence of mechanical stimuli, revealing the therapeutic potential of the combined application of BMP-2 and mechanical loads in clinical bone diseases (e.g., bone fractures and osteoporosis); however, the underlying mechanisms remain elusive. In this study, we found that cyclic stretch or BMP-2 alone increased the expression of osteoblastic differentiation markers, including alkaline phosphatase (ALP) and runt-related transcription factor 2 (Runx2), as shown by RT-qPCR, western blot analysis and ALP activity test. Furthermore, our results revealed that cyclic mechanical stretch with 10% elongation at 0.1 Hz significantly enhanced the BMP-2-induced upregulation of ALP and Runx2 expression in osteoblast-like MC3T3-E1 cells. Cyclic stretch also inhibited the BMP-2-induced upregulation of Hes-related family bHLH transcription factor with YRPW motif 1 (Hey1, measured by RT-qPCR and immunofluorescence staining), a potent negative regulator of osteogenesis. Moreover, the transient transfection of a Hey1 expression plasmid (pcDNA3.1-Hey1) significantly reversed the effects of cyclic stretch on the BMP-2-induced upregulation of differentiation markers in the MC3T3-E1 cells. This revealed the importance of Hey1 in modulating BMP-2-induced osteoblastic differentiation in response to cyclic stretch. Taken together, our results demonstrated that cyclic stretch enhanced the BMP-2‑induced osteoblastic differentiation through the inhibition of Hey1. The present study broadens our fundamental knowledge of osteoblastic mechanotransduction and also sheds new insight into the mechanisms through which the combined application of BMP-2 and mechanical load promotes osteogenesis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bone Morphogenetic Protein 2 human, BMP-2, recombinant, expressed in HEK 293 cells, HumanKine, suitable for cell culture
Sigma-Aldrich
BMP-2 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE)
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Bone Morphogenetic Protein 2 human, Carrier Free, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Bone Morphogenetic Protein 2 human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®