Merck
Measurement of Bmax and Kd with the glycine transporter 1 radiotracer ¹⁸F-MK6577 using a novel multi-infusion paradigm.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism (2015-07-23)
Yan Xia, Ming-Qiang Zheng, Daniel Holden, Shu-fei Lin, Michael Kapinos, Jim Ropchan, Jean-Dominique Gallezot, Yiyun Huang, Richard E Carson
ABSTRAKT
Glycine is a co-agonist of glutamate at the NMDA receptor. Glycine transporter 1 (GlyT1) inhibitors are reported to be potential therapeutic agents for schizophrenia. (18)F-MK6577 is a new positron emission tomography (PET) radiotracer useful for imaging brain GlyT1 and its occupancy in humans. We devised a novel multi-infusion paradigm of radiolabeled and unlabeled compound and an iterative linear/nonlinear alternating fitting method to allow for the determination of in vivo affinity (Kd) and target concentration (Bmax) images, constraining Kd to be uniform across the brain. This paradigm was tested with (18)F-MK6577 in baboons. Voxel-based analysis produced high quality Bmax images and reliable Kd estimates, and also suggested that the nondisplaceable distribution volume (VND) is not uniform throughout the brain. In vivo GlyT1 Kd was estimated to be 1.87 nmol/L for (18)F-MK6577, and the rank order of GlyT1 distribution measured in the baboon brain was: high in the brainstem (133 nmol/L), medium in the cerebellum (83 nmol/L), and low in the cortex (30 nmol/L). These in vivo Kd and Bmax values agreed well with those determined in vitro, thus validating our novel multi-infusion approach.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Tantalum(V) ethoxide, packaged for use in deposition systems
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)