Merck
  • Altered activities of kininase II, an angiotensin converting enzyme, prekallikrein, and nitric oxide in Kuwaiti patients with type 2 diabetes.
Altered activities of kininase II, an angiotensin converting enzyme, prekallikrein, and nitric oxide in Kuwaiti patients with type 2 diabetes.
International journal of immunopathology and pharmacology (2015-05-13)
J N Sharma, Kas Al-Shoumer, K M Matar, N V Madathil, Amk Al-Moalem
ABSTRAKT
The current investigation was conducted to examine kininase II or angiotensin converting enzyme (ACE), plasma prekallikrein (PK), and nitric oxide (NO) concentrations in healthy Kuwaiti subjects and newly diagnosed Kuwaiti type 2 diabetic patients before and after treatment for 6 weeks with metformin hydrochloride 500 mg twice daily after meal. With the consent of volunteers, blood and urine samples were collected after an overnight fasting. Samples were collected from the diabetic patients before and after treatment for 6 weeks. Enzyme linked immunosorbent assay (ELISA) was carried out on the aliquoted samples to measure the concentration of kininase II. NO was detected via colorimetry. Plasma Kininase II or ACE levels were significantly (P <0.01) increased by 18% in untreated diabetics when compared with healthy volunteers. However, after treatment there was a significant decrease of 20% in their ACE levels. Plasma prekallikrein levels were raised significantly (P <0.01) by 28% in diabetic patients in contrast with the control subjects and the levels were significantly reduced (P <0.0001) by 44% after treatment with metformin hydrochloride. NO levels were found to be significantly decreased in plasma by 56% and in urine by 62% in untreated diabetic patients as compared with the healthy subjects. However, when the treated diabetic patients were compared with untreated diabetics, there was an increase of 50% in plasma and 37% in urine samples. The high levels of kininase II, prekallikrein, and reduced NO may be partly responsible for the induction of renal, cardiac, and hypertensive complications associated with type 2 diabetes. Reduced NO level is an indication of endothelial dysfunction resulting in increased blood pressure. Oral anti-diabetic treatment is associated with protective effects through the reduction of kininase II (ACE), prekallikrein, and elevation of NO levels.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder