Merck
Cognitive Function, Progression of Age-related Behavioral Changes, Biomarkers, and Survival in Dogs More Than 8 Years Old.
Journal of veterinary internal medicine (2015-10-16)
T Schütt, N Toft, M Berendt
ABSTRAKT
Canine cognitive dysfunction (CCD) is an age-dependent neurodegenerative condition dominated by changes in behavioral patterns. Cohort studies investigating cognitive status in dogs are lacking. To investigate cognitive function, progression of age-related behavioral changes, survival, and possible biomarkers of CCD in aged dogs. Fifty-one dogs >8 years old; 21 with no cognitive deficits, 17 with mild cognitive impairments (MCI) and 13 with CCD. Longitudinal study. Recruitment period of 12 months and an observational period of 24 months including a baseline and 3 planned subsequent assessments. Cognitive status was determined using validated questionnaires. Plasma Aβ-peptides were quantified using commercial ELISA assays and cytokines by a validated immunoassay. Signs characterizing dogs with CCD were aimless wandering, staring into space, avoid getting patted, difficulty finding dropped food and anxiety. Thirty-three percent of dogs with a normal cognitive status progressed to MCI and 22% classified as MCI progressed to CCD during the study period. For 6 dogs diagnosed with CCD, signs of cognitive dysfunction increased with time. A diagnosis of CCD did not affect survival. The level of plasma Aβ42 was significantly increased (P < .05) in the CCD group (92.8 ± 24.0 pg/mL) compared to the MCI (77.0 ± 12.3 pg/mL) and normal group (74.9 ± 10.0 pg/mL), but no significant differences in concentrations of systemic inflammatory markers were detected. Canine cognitive dysfunction is a progressive disorder with an individual variability in the rate of cognitive decline and clinical signs. Plasma Aβ42 seems to be an interesting plasma biomarker of CCD.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets