Merck
Junction-mediating and regulatory protein (JMY) is essential for early porcine embryonic development.
The Journal of reproduction and development (2015-06-09)
Zi Li Lin, Xiang-Shun Cui, Suk Namgoong, Nam-Hyung Kim
ABSTRAKT
Junction-mediating and regulatory protein (JMY) is a regulator of both transcription and actin filament assembly. JMY is a critical nucleation-promoting factor (NPF); however, its role in the development of mammalian embryos is poorly understood. In the current study, we investigated the functional roles of the NPF JMY in porcine embryos. Porcine embryos expressed JMY mRNA and protein, and JMY protein moved from the cytoplasm to the nucleus at later embryonic developmental stages. Knockdown of JMY by RNA interference markedly decreased the rate of blastocyst development, validating its role in the development of porcine embryos. Furthermore, injection of JMY dsRNA also impaired actin and Arp2 expression, and co-injection of actin and Arp2 mRNA partially rescued blastocyst development. Taken together, our results show that the NPF JMY is involved in the development of porcine embryos by regulating the NPF-Arp2-actin pathway.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications