Skip to Content
MilliporeSigma

860161P

Avanti

Cer 1 (d18:1(D7)/28:0/18:2)

new

Avanti Research - A Croda Brand

Synonym(s):

N-[28-linoleoyloxy octacosanoyl]-D-erythro-sphingosine(d7)

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

Empirical Formula (Hill Notation):
C64H114D7NO5
Molecular Weight:
991.69
UNSPSC Code:
12352211

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

synthetic

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (860161P-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])CCCCCCCCCC\C=C\[C@@H](O)[C@H](CO)NC(=O)CCCCCCCCCCCCCCCCCCCCCCCCCCCOC(=O)CCCCCCC\C=C/C\C=C/CCCCC

General description

Ceramide 1 (d18:1(d7)/28:0/18:2) is natural component of human skin. It plays a key role in preventing water loss, healing the skin, and maintaining the integrity of the skin.

Features and Benefits

This product is deuterated which allows for better understanding of the structure, dynamics, and function of cell membranes.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service