Sign In to View Organizational & Contract Pricing.
Select a Size
Product BDPH9BCD4E8A is currently not available for purchase. Contact Technical Service
About This Item
Skip To
Inchi Code
1S/C6H6F2N2/c7-3-1-5(9)6(10)2-4(3)8/h1-2H,9-10H2
Inchi Key
PPWRHKISAQTCCG-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Keep in dark place, inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Powder or crystals
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
